missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min

Catalog No. AA1054802
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1054802 0.5 g
AA1054804 2 g
AA1054809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1054802 Supplier Thermo Scientific Chemicals Supplier No. 01054802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrosilation, isomerization, carbonylation | CAS: 14221-01-3 | C72H60P4Pd | 1155.59 g/mol

Widely used as a catalyst for palladium-catalyzed coupling reactions including Suzuki coupling, Sonogashira coupling, Heck reaction, Negishi coupling and Stille coupling. Used in isomerization, carbonylation, oxidation, and C-C bond formation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14221-01-3
Molecular Formula C72H60P4Pd
Molecular Weight (g/mol) 1155.59
MDL Number MFCD00010012
InChI Key NFHFRUOZVGFOOS-UHFFFAOYSA-N
PubChem CID 11979704
SMILES [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 100°C to 105°C
Assay Percent Range 99.8% (metals basis), Pd 9% min
Quantity 0.5 g
Beilstein 6704828
Sensitivity Air sensitive
Synonym tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
Solubility Information Insoluble in water. Soluble in benzene,ethanol and chloroform
IUPAC Name palladium;triphenylphosphane
Formula Weight 1155.57
Physical Form Crystalline
Chemical Name or Material Tetrakis(triphenylphosphine)palladium
Show More Show Less
Hazard Category H302-H500
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 238-086-9
TSCA Yes
storageNote1 Air Sensitive
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only