missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)nickel(0), 95%

Catalog No. AC223980050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223980050 5 g Glass bottle
AC223980010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223980050 Supplier Thermo Scientific Chemicals Supplier No. 223980050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15133-82-1 | C72H60NiP4 | 1107.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15133-82-1
Molecular Formula C72H60NiP4
Molecular Weight (g/mol) 1107.86
MDL Number MFCD00010011
InChI Key KFBKRCXOTTUAFS-UHFFFAOYSA-N
Synonym ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx.
PubChem CID 11979963
IUPAC Name nickel;triphenylphosphane
SMILES [Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 122.0°C to 124.0°C
Color Brown to Brown-Rust
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula [(C6H5)3P]4Ni
Quantity 5 g
Fieser 06,570; 07,357; 09,450; 12,467; 13,294; 16,317
Formula Weight 1107.89
Percent Purity 95%
Physical Form Fine Powder
Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0), 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
Flammable solid.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

RUO – Research Use Only