missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(hydroxymethyl)phosphonium Sulfate (ca. 70-80% in Water), TCI America™

Catalog No. T108925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T108925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T108925G Supplier TCI America Supplier No. T108925G
Only null left

Chemical Identifiers

CAS 55566-30-8
Molecular Formula C8H24O12P2S
Molecular Weight (g/mol) 406.276
MDL Number MFCD00054243
InChI Key YIEDHPBKGZGLIK-UHFFFAOYSA-L
PubChem CID 41478
IUPAC Name tetrakis(hydroxymethyl)phosphanium;sulfate
SMILES C(O)[P+](CO)(CO)CO.C(O)[P+](CO)(CO)CO.[O-]S(=O)(=O)[O-]

Specifications

Color Colorless
Quantity 25 g
UN Number 2922
Formula Weight 406.28
Physical Form Liquid
Chemical Name or Material Tetrakis(hydroxymethyl)phosphonium Sulfate (ca. 70-80% in Water)
EINECSNumber (2)-2003
RTECSNumber TA2570000
TSCA Yes