accessibility menu, dialog, popup

UserName

OHPHOHOHOOHPHOHOHOOSOOO

CAS RN 55566-30-8

IUPAC Name: bis(tetrakis(hydroxymethyl)phosphanium) sulfate
Synonyms: tetrakis(hydroxymethyl)phosphonium sulfate
Molecular Weight (g/mol): 406.28
Molecular Formula: C8H24O12P2S
InChi Key: YIEDHPBKGZGLIK-UHFFFAOYSA-L
SMILES: [O-]S([O-])(=O)=O.OC[P+](CO)(CO)CO.OC[P+](CO)(CO)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results