missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(dimethylamino)diboron, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 468540250
Specifications
| Tetrakis(dimethylamino)diboron | |
| -33°C | |
| 55°C to 57°C (2.5 mmHg) | |
| Glass Bottle | |
| 1.4680 to 1.4760 (20°C, 589 nm) | |
| 25 g | |
| CN(C)B(B(N(C)C)N(C)C)N(C)C | |
| 197.93 | |
| Liquid |
| 1630-79-1 | |
| 0.926 | |
| Conforms to structure | |
| C8H24B2N4 | |
| MFCD00048012 | |
| KMCDRSZVZMXKRL-UHFFFAOYSA-N | |
| dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine | |
| 197.93 |
Chemical Identifiers
| 1630-79-1 | |
| 197.93 | |
| KMCDRSZVZMXKRL-UHFFFAOYSA-N | |
| CN(C)B(B(N(C)C)N(C)C)N(C)C |
| C8H24B2N4 | |
| MFCD00048012 | |
| dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine |
Safety and Handling
GHS08: Health hazard
GHS07: Exclamation mark
ShelfLife : 2 years
Recommended Storage : Refrigerator +4°C