missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)diboron 95.0+%, TCI America™

Catalog No. T30191G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30191G Supplier TCI America Supplier No. T30191G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1630-79-1
Molecular Formula C8H24B2N4
Molecular Weight (g/mol) 197.93
MDL Number MFCD00048012
InChI Key KMCDRSZVZMXKRL-UHFFFAOYSA-N
Synonym Tetrakis(dimethylamino)diborane
PubChem CID 137125
IUPAC Name dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine
SMILES CN(C)B(B(N(C)C)N(C)C)N(C)C

Specifications

Melting Point -33°C
Color Yellow
Boiling Point 87°C
Quantity 1 g
Formula Weight 197.93
Percent Purity ≥95.0% (N)
Physical Form Liquid
Chemical Name or Material Tetrakis(dimethylamino)diboron
TSCA No
Recommended Storage Refrigerator