missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(decyl)ammonium bromide, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Melting Point | 88.0°C |
| Color | White |
| Physical Form | Fine Powder |
| Quantity | 10 g |
| Assay Percent Range | 97.5% min. (Argentometry) |
| Formula Weight | 659.01 |
| Percent Purity | 98% |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Chemical Name or Material | Tetrakis(decyl)ammonium bromide |
Safety and Handling
| EINECSNumber | 239-013-3 |
RUO – Research Use Only