accessibility menu, dialog, popup

UserName

CH3NCH3CH3CH3Br

CAS RN 14937-42-9

IUPAC Name: tetrakis(decyl)azanium bromide
Synonyms: tetrakis(decyl)azanium bromide
Molecular Weight (g/mol): 659.02
Molecular Formula: C40H84BrN
InChi Key: AHNISXOXSNAHBZ-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)