missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(acetonitrile)copper(I) Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7137343
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26661G 1 g
T26665G 5 g
T266625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T26661G Supplier TCI America Supplier No. T26661G
Only null left

Chemical Identifiers

CAS 15418-29-8
Molecular Formula C8H12BCuF4N4
Molecular Weight (g/mol) 314.56
MDL Number MFCD09265110
InChI Key YZGSKMIIVMCEFE-UHFFFAOYSA-N
Synonym tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate
PubChem CID 10990673
IUPAC Name λ¹-copper(1+) tetrakis(acetonitrile) tetrafluoroboranuide
SMILES [Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F

Specifications

Melting Point 164°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 314.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(acetonitrile)copper(I) Tetrafluoroborate