accessibility menu, dialog, popup

UserName

CAS RN 15418-29-8

IUPAC Name: λ¹-copper(1+) tetrakis(acetonitrile) tetrafluoroboranuide
Synonyms: λ¹-copper(1+) tetrakis(acetonitrile) tetrafluoroborate
Molecular Weight (g/mol): 314.56
Molecular Formula: C8H12BCuF4N4
InChi Key: YZGSKMIIVMCEFE-UHFFFAOYSA-N
SMILES: [Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)