missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(4-hydroxyphenyl)ethylene 97.0+%, TCI America™
$523.79 - $2095.21
Chemical Identifiers
| CAS | 119301-59-6 |
|---|---|
| Molecular Formula | C26H20O4 |
| Molecular Weight (g/mol) | 396.442 |
| InChI Key | QQUZHNPGWNIYMK-UHFFFAOYSA-N |
| PubChem CID | 10200771 |
| IUPAC Name | 4-[1,2,2-tris(4-hydroxyphenyl)ethenyl]phenol |
| SMILES | C1=CC(=CC=C1C(=C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C4=CC=C(C=C4)O)O |
Chemical Identifiers
| 119301-59-6 | |
| 396.442 | |
| 10200771 | |
| C1=CC(=CC=C1C(=C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C4=CC=C(C=C4)O)O |
| C26H20O4 | |
| QQUZHNPGWNIYMK-UHFFFAOYSA-N | |
| 4-[1,2,2-tris(4-hydroxyphenyl)ethenyl]phenol |
Specifications
| 119301-59-6 | |
| C26H20O4 | |
| QQUZHNPGWNIYMK-UHFFFAOYSA-N | |
| 4-[1,2,2-tris(4-hydroxyphenyl)ethenyl]phenol | |
| 10200771 | |
| ≥97.0% (GC) | |
| Tetrakis(4-hydroxyphenyl)ethylene |
| 323°C | |
| 1 g | |
| C1=CC(=CC=C1C(=C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C4=CC=C(C=C4)O)O | |
| 396.442 | |
| 396.44 | |
| Crystalline Powder |
Safety and Handling
TSCA : No