missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(4-hydroxyphenyl)ethylene 97.0+%, TCI America™

Catalog No. T31421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31421G 1 g
T31425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31421G Supplier TCI America Supplier No. T31421G
Only null left

Chemical Identifiers

CAS 119301-59-6
Molecular Formula C26H20O4
Molecular Weight (g/mol) 396.442
InChI Key QQUZHNPGWNIYMK-UHFFFAOYSA-N
PubChem CID 10200771
IUPAC Name 4-[1,2,2-tris(4-hydroxyphenyl)ethenyl]phenol
SMILES C1=CC(=CC=C1C(=C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C4=CC=C(C=C4)O)O

Specifications

Melting Point 323°C
Quantity 1 g
Formula Weight 396.44
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(4-hydroxyphenyl)ethylene