missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(4-aminophenyl)methane 95.0+%, TCI America™

Catalog No. p-7137493
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
T2947200MG 200 mg
T29471G 1 g
2 options

Catalog No. T2947200MG

Supplier: TCI America T2947200MG

Only null left

Chemical Identifiers

CAS 60532-63-0
Molecular Formula C25H24N4
Molecular Weight (g/mol) 380.50
MDL Number MFCD12546930
InChI Key LNHGLSRCOBIHNV-UHFFFAOYSA-N
Synonym 4,4′,4′′,4′′′-Methanetetrayltetraaniline
PubChem CID 11825069
IUPAC Name 4-[tris(4-aminophenyl)methyl]aniline
SMILES NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Melting Point 320°C
Color White-Yellow
Quantity 200 mg
Formula Weight 380.50
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(4-aminophenyl)methane