CAS RN 60532-63-0
IUPAC Name:
4-[tris(4-aminophenyl)methyl]aniline
Synonyms:
4-[tris(4-aminophenyl)methyl]aniline
Molecular Weight (g/mol):
380.5
Molecular Formula:
C25H24N4
InChi Key:
LNHGLSRCOBIHNV-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
1
–
1
of
1
results
1
Tetrakis(4-aminophenyl)methane 95.0+%, TCI America™
CAS: 60532-63-0 Molecular Formula: C25H24N4 Molecular Weight (g/mol): 380.50 MDL Number: MFCD12546930 InChI Key: LNHGLSRCOBIHNV-UHFFFAOYSA-N Synonym: 4,4′,4′′,4′′′-Methanetetrayltetraaniline PubChem CID: 11825069 IUPAC Name: 4-[tris(4-aminophenyl)methyl]aniline SMILES: NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1