missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(4-aminophenyl)methane 95.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 320°C |
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 380.50 |
| Percent Purity | ≥95.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tetrakis(4-aminophenyl)methane |
Sécurité et manipulation
| TSCA | No |