missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(2,4-pentanedionato)titanium(IV) (ca. 63% in Isopropyl Alcohol), TCI America™
$63.38 - $363.49
Chemical Identifiers
| CAS | 17501-79-0 |
|---|---|
| Molecular Formula | C20H32O8Ti |
| Molecular Weight (g/mol) | 448.335 |
| MDL Number | MFCD00045004 |
| InChI Key | RYSXWUYLAWPLES-MTOQALJVSA-N |
| Synonym | Acetylacetone Titanium(IV) Salt, Titanium(IV) Acetylacetonate |
| PubChem CID | 44629996 |
| IUPAC Name | (Z)-4-hydroxypent-3-en-2-one;titanium |
| SMILES | CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Ti] |
Chemical Identifiers
| 17501-79-0 | |
| 448.335 | |
| RYSXWUYLAWPLES-MTOQALJVSA-N | |
| 44629996 | |
| CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Ti] |
| C20H32O8Ti | |
| MFCD00045004 | |
| Acetylacetone Titanium(IV) Salt, Titanium(IV) Acetylacetonate | |
| (Z)-4-hydroxypent-3-en-2-one;titanium |
Specifications
| 17501-79-0 | |
| MFCD00045004 | |
| 1219 | |
| RYSXWUYLAWPLES-MTOQALJVSA-N | |
| (Z)-4-hydroxypent-3-en-2-one;titanium | |
| 44629996 | |
| Liquid |
| C20H32O8Ti | |
| 25 g | |
| Acetylacetone Titanium(IV) Salt, Titanium(IV) Acetylacetonate | |
| CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Ti] | |
| 448.335 | |
| 444.30 | |
| Tetrakis(2,4-pentanedionato)titanium(IV) (ca. 63% in Isopropyl Alcohol) |
Safety and Handling
TSCA : Yes