accessibility menu, dialog, popup

UserName

CAS RN 17501-79-0

TiCH3OCHCH3OCH3OCHCH3OCH3OCHCH3OCH3OCHCH3O

CAS RN 17501-79-0

IUPAC Name: titanium(4+) tetrakis(2,4-dioxopentan-3-ide)
Synonyms: titanium(4+) tetraacetylacetonate
Molecular Weight (g/mol): 444.3
Molecular Formula: C20H28O8Ti
InChi Key: QMJUQLJXRRGCJS-UHFFFAOYSA-N
SMILES: [Ti+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results