missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV), Thermo Scientific™
$165.95 - $165.95
Chemical Identifiers
| CAS | 41706-15-4 |
|---|---|
| Molecular Formula | C44H80NbO8 |
| Molecular Weight (g/mol) | 830.022 |
| MDL Number | MFCD00145379 |
| InChI Key | WYTIGJVEVROCSD-DSORRXEFSA-N |
| Synonym | tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato niobium iv, nb tmhd 4, niobium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, 4e-5-bis 3e-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy niobio oxy-2,2,6,6-tetramethylhept-4-en-3-one |
| PubChem CID | 72376395 |
| IUPAC Name | (Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;niobium |
| SMILES | CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Nb] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 41706-15-4 | |
| 830.022 | |
| WYTIGJVEVROCSD-DSORRXEFSA-N | |
| 72376395 | |
| CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Nb] |
| C44H80NbO8 | |
| MFCD00145379 | |
| tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato niobium iv, nb tmhd 4, niobium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, 4e-5-bis 3e-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy niobio oxy-2,2,6,6-tetramethylhept-4-en-3-one | |
| (Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;niobium |
Specifications
| 41706-15-4 | |
| 325°C (decomposition) | |
| MFCD00145379 | |
| tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato niobium iv, nb tmhd 4, niobium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, 4e-5-bis 3e-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy niobio oxy-2,2,6,6-tetramethylhept-4-en-3-one | |
| CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Nb] | |
| 830.022 | |
| 826 | |
| Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV) |
| 219°C to 220°C | |
| C44H80NbO8 | |
| 1 g | |
| WYTIGJVEVROCSD-DSORRXEFSA-N | |
| (Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;niobium | |
| 72376395 | |
| Powder |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures