accessibility menu, dialog, popup

UserName

CAS RN 41706-15-4

IUPAC Name: tetrakis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) niobium
Synonyms: tetrakis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) niobium
Molecular Weight (g/mol): 830.02
Molecular Formula: C44H80NbO8
InChi Key: WYTIGJVEVROCSD-UHFFFAOYSA-N
SMILES: [Nb].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results