missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraisopropylthiuram Disulfide 98.0+%, TCI America™
Supplier: TCI America T155525G
Specifications
| Tetraisopropylthiuram Disulfide | |
| 115°C | |
| C14H28N2S4 | |
| 25 g | |
| ZUYREEAWHZRZDX-UHFFFAOYSA-N | |
| N,N-bis(propan-2-yl){[bis(propan-2-yl)carbamothioyl]disulfanyl}carbothioamide | |
| 95876 | |
| ≥98.0% (N) |
| 4136-91-8 | |
| Yellow | |
| MFCD00009860 | |
| Bis(diisopropylthiocarbamoyl) Disulfide | |
| CC(C)N(C(C)C)C(=S)SSC(=S)N(C(C)C)C(C)C | |
| 352.63 | |
| 352.63 | |
| Crystalline Powder |
Chemical Identifiers
| 4136-91-8 | |
| 352.63 | |
| ZUYREEAWHZRZDX-UHFFFAOYSA-N | |
| 95876 | |
| CC(C)N(C(C)C)C(=S)SSC(=S)N(C(C)C)C(C)C |
| C14H28N2S4 | |
| MFCD00009860 | |
| Bis(diisopropylthiocarbamoyl) Disulfide | |
| N,N-bis(propan-2-yl){[bis(propan-2-yl)carbamothioyl]disulfanyl}carbothioamide |
Safety and Handling
TSCA : No