missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraisopropylthiuram Disulfide 98.0+%, TCI America™
$233.00 - $1151.98
Chemical Identifiers
| CAS | 4136-91-8 |
|---|---|
| Molecular Formula | C14H28N2S4 |
| Molecular Weight (g/mol) | 352.63 |
| MDL Number | MFCD00009860 |
| InChI Key | ZUYREEAWHZRZDX-UHFFFAOYSA-N |
| Synonym | Bis(diisopropylthiocarbamoyl) Disulfide |
| PubChem CID | 95876 |
| IUPAC Name | N,N-bis(propan-2-yl){[bis(propan-2-yl)carbamothioyl]disulfanyl}carbothioamide |
| SMILES | CC(C)N(C(C)C)C(=S)SSC(=S)N(C(C)C)C(C)C |
Chemical Identifiers
| 4136-91-8 | |
| 352.63 | |
| ZUYREEAWHZRZDX-UHFFFAOYSA-N | |
| 95876 | |
| CC(C)N(C(C)C)C(=S)SSC(=S)N(C(C)C)C(C)C |
| C14H28N2S4 | |
| MFCD00009860 | |
| Bis(diisopropylthiocarbamoyl) Disulfide | |
| N,N-bis(propan-2-yl){[bis(propan-2-yl)carbamothioyl]disulfanyl}carbothioamide |
Specifications
| 4136-91-8 | |
| Yellow | |
| MFCD00009860 | |
| Bis(diisopropylthiocarbamoyl) Disulfide | |
| CC(C)N(C(C)C)C(=S)SSC(=S)N(C(C)C)C(C)C | |
| 352.63 | |
| 352.63 | |
| Crystalline Powder |
| 115°C | |
| C14H28N2S4 | |
| 5 g | |
| ZUYREEAWHZRZDX-UHFFFAOYSA-N | |
| N,N-bis(propan-2-yl){[bis(propan-2-yl)carbamothioyl]disulfanyl}carbothioamide | |
| 95876 | |
| ≥98.0% (N) | |
| Tetraisopropylthiuram Disulfide |
Safety and Handling
TSCA : No