missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraisopropyl Orthosilicate 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 185°C |
| Quantity | 10 mL |
| UN Number | 1760 |
| Formula Weight | 264.44 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tetraisopropyl Orthosilicate |
Safety and Handling
| EINECSNumber | (2)-2048 |
| TSCA | Yes |