accessibility menu, dialog, popup

UserName

CH3H3COSiOCH3CH3OH3CCH3OCH3H3C

CAS RN 1992-48-9

IUPAC Name: tetrakis(propan-2-yl) silicate
Synonyms: tetraisopropyl silicate
Molecular Weight (g/mol): 264.44
Molecular Formula: C12H28O4Si
InChi Key: ZUEKXCXHTXJYAR-UHFFFAOYSA-N
SMILES: CC(C)O[Si](OC(C)C)(OC(C)C)OC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results