missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrothiopyran-4-one 1,1-Dioxide 98.0+%, TCI America™
Supplier: TCI America T33201G
Specifications
| Tetrahydrothiopyran-4-one 1,1-Dioxide | |
| 172°C | |
| C5H8O3S | |
| 1 g | |
| XFMQGQAAHOGFQS-UHFFFAOYSA-N | |
| 1,1-dioxothian-4-one | |
| 262230 | |
| ≥98.0% (GC) |
| 17396-35-9 | |
| White-Yellow | |
| MFCD00460698 | |
| 4-Oxothiane 1,1-Dioxide | |
| C1CS(=O)(=O)CCC1=O | |
| 148.176 | |
| 148.18 | |
| Crystalline Powder |
Chemical Identifiers
| 17396-35-9 | |
| 148.176 | |
| XFMQGQAAHOGFQS-UHFFFAOYSA-N | |
| 262230 | |
| C1CS(=O)(=O)CCC1=O |
| C5H8O3S | |
| MFCD00460698 | |
| 4-Oxothiane 1,1-Dioxide | |
| 1,1-dioxothian-4-one |
Safety and Handling
TSCA : No