accessibility menu, dialog, popup

UserName

CAS RN 17396-35-9

IUPAC Name: 1λ⁶-thiane-1,1,4-trione
Synonyms: 1λ⁶-thiane-1,1,4-trione
Molecular Weight (g/mol): 148.18
Molecular Formula: C5H8O3S
InChi Key: XFMQGQAAHOGFQS-UHFFFAOYSA-N
SMILES: O=C1CCS(=O)(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results