missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran-4-one 1,1-Dioxide 98.0+%, TCI America™

Catalog No. T3320200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3320200MG 200 mg
T33201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3320200MG Supplier TCI America Supplier No. T3320200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17396-35-9
Molecular Formula C5H8O3S
Molecular Weight (g/mol) 148.176
MDL Number MFCD00460698
InChI Key XFMQGQAAHOGFQS-UHFFFAOYSA-N
Synonym 4-Oxothiane 1,1-Dioxide
PubChem CID 262230
IUPAC Name 1,1-dioxothian-4-one
SMILES C1CS(=O)(=O)CCC1=O

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 200 mg
Formula Weight 148.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahydrothiopyran-4-one 1,1-Dioxide