missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophene, 98%

Catalog No. AAL1390836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1390836 500 g
AAL1390822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1390836 Supplier Thermo Scientific Chemicals Supplier No. L1390836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-01-0 | C4H8S | 88.17 g/mol

Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.

Solubility
Immiscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 110-01-0
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00005476
InChI Key RAOIDOHSFRTOEL-UHFFFAOYSA-N
Synonym tetrahydrothiophene, thiophene, tetrahydro, thiophane, thiacyclopentane, tetramethylene sulfide, thilane, tetrahydrothiophen, pennodorant 1013, thiofan, thiolan
PubChem CID 1127
ChEBI CHEBI:48458
IUPAC Name thiolane
SMILES C1CCSC1

Specifications

Melting Point -96°C
Density 0.999
Boiling Point 120°C to 121°C
Flash Point 19°C (66°F)
Refractive Index 1.504
Quantity 500 g
UN Number UN2412
Beilstein 102392
Merck Index 14,9218
Solubility Information Immiscible in water.
Formula Weight 88.17
Percent Purity 98%
Chemical Name or Material Tetrahydrothiophene
Show More Show Less
Hazard Category H225-H302+H312-H315-H319
Hazard Statement GHS H Statement
H225-H302-H312-H332-H315-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROTHIOPHENE
EINECSNumber 203-728-9
RTECSNumber XN0370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only