missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophene 1-oxide, 97%

Catalog No. AAA1750209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1750209 10 g
AAA1750218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1750209 Supplier Thermo Scientific Chemicals Supplier No. A1750209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1600-44-8 | C4H8OS | 104.167 g/mol

In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

Solubility
Fully miscible in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1600-44-8
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00005477
InChI Key ISXOBTBCNRIIQO-UHFFFAOYSA-N
Synonym tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide
PubChem CID 1128
IUPAC Name thiolane 1-oxide
SMILES C1CCS(=O)C1

Specifications

Density 1.173
Boiling Point 235°C to 237°C
Flash Point 112°C (233°F)
Refractive Index 1.52
Quantity 10 g
Beilstein 105274
Solubility Information Fully miscible in water.
Formula Weight 104.17
Percent Purity 97%
Chemical Name or Material Tetrahydrothiophene 1-oxide
EINECSNumber 216-493-2
RTECSNumber XN0830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only