Learn More
Tetrahydropyran-4-ylacetic acid, 97%, Thermo Scientific™
$126.82 - $126.82
Chemical Identifiers
| CAS | 85064-61-5 |
|---|---|
| Molecular Formula | C7H11O3 |
| Molecular Weight (g/mol) | 143.16 |
| MDL Number | MFCD01631204 |
| InChI Key | PBXYNWPYMVWJAH-UHFFFAOYSA-M |
| Synonym | tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid |
| PubChem CID | 2773575 |
| IUPAC Name | 2-(oxan-4-yl)acetic acid |
| SMILES | [O-]C(=O)CC1CCOCC1 |
Chemical Identifiers
| 85064-61-5 | |
| 143.16 | |
| PBXYNWPYMVWJAH-UHFFFAOYSA-M | |
| 2773575 | |
| [O-]C(=O)CC1CCOCC1 |
| C7H11O3 | |
| MFCD01631204 | |
| tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid | |
| 2-(oxan-4-yl)acetic acid |
Specifications
| 85064-61-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD01631204 | |
| tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid | |
| [O-]C(=O)CC1CCOCC1 | |
| 143.16 | |
| 144.17 | |
| Tetrahydropyran-4-ylacetic acid |
| 96.5 | |
| 97% | |
| C7H11O3 | |
| 1 g | |
| PBXYNWPYMVWJAH-UHFFFAOYSA-M | |
| 2-(oxan-4-yl)acetic acid | |
| 2773575 | |
| 97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.