CAS RN 85064-61-5
IUPAC Name:
2-(oxan-4-yl)acetate
Synonyms:
2-(oxan-4-yl)acetate
Molecular Weight (g/mol):
143.16
Molecular Formula:
C7H11O3
InChi Key:
PBXYNWPYMVWJAH-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CC1CCOCC1
1
–
2
of
2
results
1
Tetrahydropyran-4-ylacetic acid, 97%, Thermo Scientific™
CAS: 85064-61-5 Molecular Formula: C7H11O3 Molecular Weight (g/mol): 143.16 MDL Number: MFCD01631204 InChI Key: PBXYNWPYMVWJAH-UHFFFAOYSA-M Synonym: tetrahydropyranyl-4-acetic acid,tetrahydropyran-4-yl-acetic acid,2-oxan-4-yl acetic acid,2-tetrahydro-2h-pyran-4-yl acetic acid,tetrahydropyran-4-ylacetic acid,oxan-4-ylacetic acid,tetrahydro-2h-pyran-4-ylacetic acid,2h-pyran-4-acetic acid, tetrahydro,2-4-tetrahydropyranyl acetic acid,tetrahydropyran-4-acetic acid PubChem CID: 2773575 IUPAC Name: 2-(oxan-4-yl)acetic acid SMILES: [O-]C(=O)CC1CCOCC1