missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropalmatine, 98%

Catalog No. AAJ6332806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6332806 5 g
AAJ6332803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6332806 Supplier Thermo Scientific Chemicals Supplier No. J6332806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2934-97-6 | C21H25NO4 | 355.434 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2934-97-6
Molecular Formula C21H25NO4
Molecular Weight (g/mol) 355.434
MDL Number MFCD00214191
InChI Key AEQDJSLRWYMAQI-UHFFFAOYSA-N
Synonym tetrahydropalmatine, rotundine, tetrahydropalmatin, rotundinum, 2,3,9,10-tetramethoxy-5,8,13,13a-tetrahydro-6h-isoquino 3,2-a isoquinoline, corydalis b, 6h-dibenzo a,g quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy, gnf-pf-3943, tetrahydropalmatine hydrochloride, 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5h-isoquinolino 2,1-b isoquinoline
PubChem CID 5417
IUPAC Name 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[2,1-b]isoquinoline
SMILES COC1=C(C2=C(CC3C4=CC(=C(C=C4CCN3C2)OC)OC)C=C1)OC

Specifications

Melting Point 155°C
Color Yellow
Quantity 5 g
Formula Weight 355.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tetrahydropalmatine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only