missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl Butyrate 98.0+%, TCI America™

Catalog No. T0878500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0878500ML 500 mL
T087825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0878500ML Supplier TCI America Supplier No. T0878500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2217-33-6
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00059734
InChI Key DPZVDLFOAZNCBU-UHFFFAOYSA-N
Synonym Butyric Acid Tetrahydrofurfuryl Ester, 2-(Butyryloxymethyl)tetrahydrofuran
PubChem CID 551316
IUPAC Name oxolan-2-ylmethyl butanoate
SMILES CCCC(=O)OCC1CCCO1

Specifications

Color Yellow
Quantity 500 mL
Physical Form Clear Liquid at 20°C
Boiling Point 227°C
Formula Weight 172.22
Percent Purity ≥98.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Butyrate