accessibility menu, dialog, popup

UserName

CAS RN 2217-33-6

IUPAC Name: (oxolan-2-yl)methyl butanoate
Synonyms: oxolan-2-ylmethyl butanoate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: DPZVDLFOAZNCBU-UHFFFAOYNA-N
SMILES: CCCC(=O)OCC1CCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)