missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Tetrahydrofurfuryl alcohol, 98%

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol

$53.32 - $190.66

Chemical Identifiers

CAS 97-99-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00005372
InChI Key BSYVTEYKTMYBMK-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
PubChem CID 7360
SMILES OCC1CCCO1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA17078AE
View Documents
Thermo Scientific Chemicals
A17078AE
100 mL
Each for $53.32
Only null left
 
AAA17078AP
View Documents
Thermo Scientific Chemicals
A17078AP
500 mL
Each for $74.86
Only null left
 
AAA170780F
View Documents
Thermo Scientific Chemicals
A170780F
2500 mL
Each for $190.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

97-99-4
102.13
BSYVTEYKTMYBMK-UHFFFAOYNA-N
7360
C5H10O2
MFCD00005372
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
OCC1CCCO1

Specifications

<-80°C
1.053
100 mL
C5H10O2
102723
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
OCC1CCCO1
102.13
102.13
(+/-)-Tetrahydrofurfuryl alcohol
97-99-4
178°C
1.452
MFCD00005372
14,9213
BSYVTEYKTMYBMK-UHFFFAOYNA-N
(oxolan-2-yl)methanol
7360
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.

P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c

H227-H319-H335-H360Df

EINECSNumber : 202-625-6

RTECSNumber : LU2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only