missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Tetrahydrofurfuryl alcohol, 98%

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol

Supplier:  Thermo Scientific Chemicals A17078AP

 View more versions of this product

Catalog No. AAA17078AP


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+/-)-Tetrahydrofurfuryl alcohol
97-99-4
178°C
1.452
MFCD00005372
14,9213
BSYVTEYKTMYBMK-UHFFFAOYNA-N
oxolan-2-ylmethanol
7360
98%
<-80°C
1.053
500 mL
C5H10O2
102723
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
OCC1CCCO1
102.13
102.13

Chemical Identifiers

97-99-4
102.13
BSYVTEYKTMYBMK-UHFFFAOYNA-N
7360
C5H10O2
MFCD00005372
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
OCC1CCCO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.

P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c

H227-H319-H335-H360Df

EINECSNumber : 202-625-6

RTECSNumber : LU2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only