missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Stabilized with BHT, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical

Catalog No. p-7158371
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607328 1 L Amber Glass Bottle
18607329 4 L Amber Glass Bottle
18607331 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607328 Supplier Spectrum Chemical Mfg Cor Supplier No. HP8321LT
Only null left

HP832, 109-99-9, C4H8O

Spectrum™ Chemical Tetrahydrofuran, Stabilized with BHT, Exceeds A.C.S. Specifications, HPLC Grade , is a colorless, organic liquid with a low viscosity that is water miscible. It has the ability, as an aprotic solvent, to dissolve both polar and non-polar chemical compounds. All HPLC and Ultra-pure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

CAS Max % 1
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 0.0003
Water 0.0003
Boiling Point 65°C to 66°C
Percent Purity 99.9%
Grade ACS/HPLC
Chemical Name or Material Tetrahydrofuran