missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Stabilized with 0.025% BHT, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7166627
Change view
Click to view available options
Quantity:
100 mL
120 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614358 100 mL Amber Glass Bottle
18614359 120 mL Amber Glass Bottle
18614360 500 mL Amber Glass Bottle
18614361 1 L Amber Glass Bottle
18614362 4 L Amber Glass Bottle
18614363 20 L Steel Can
18614364 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18614358 Supplier Spectrum Chemical Mfg Cor Supplier No. T1025100ML
Only null left

Chemical Identifiers

CAS 109-99-9
Molecular Weight (g/mol) 72.11

Specifications

CAS Max % 1
Quantity 100 mL
Identification To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 0.0003
Water 0.0005
Boiling Point 65°C to 66°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material Tetrahydrofuran

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.