missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, 99.5+%, for spectroscopy

CAS: 109-99-9 | C4H8O | 72.11 g/mol

Supplier:  Thermo Scientific Chemicals 222160010

 View more versions of this product

Catalog No. AC222160010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Specifications

Tetrahydrofuran
For spectroscopy
1 max. at 215nm, 0.5 max. at 230nm, 0.26 max. at 240nm, 0.1 max. at 250nm, 0.05 max. at 260nm, 0.04 max. at 270nm, 0.02 max. at 280nm, (1 cm cell vs HPLC-grade water), (1 cm cell vs HPLC-grade water) at 0nm
≤0.0006meq/g
66.0°C
−21°C
Glass Bottle
1.407
99.5+%
C4H8O
Vapor Pressure: 200mbar at 20°C
≤0.03%
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
Spectroscopy
-108.4°C
109-99-9,128-37-0
≤0.0002meq/g
0.8800g/mL
1 L
Pass Test
Liquid
≤0.005%
0.88
MFCD00005356
0.55 mPa.s (20°C)
01,1140; 02,398; 03,278; 05,649; 06,570
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
C1CCOC1
72.11
CHEBI:26911
≥99.5%

Chemical Identifiers

109-99-9
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only