missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, ReagentPlus™, ≥99.0%, Solstice

Catalog No. 60044963
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044963 20 L Stainless Steel Can
60044961 1 L Glass Bottle
60044962 2.5 L Glass Bottle
60044960 10 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60044963 Supplier Solstice Supplier No. 17881020L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Grade, contains 250 ppm BHT as inhibitor,≥99.0%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Color Colorless
Density 0.889g/mL at 25°C
Quantity 20 L
Explosion Limit 0.118
Flash Point −17°C (1.4°F)
Ignition Point 610°F
Packaging Stainless Steel Can
Physical Form Liquid
Refractive Index n20/D 1.407(lit.)
Vapor Pressure 114mmHg (15°C), 143mmHg (20°C)
UN Number UN2056
Beilstein 102391
Boiling Point 65°C
Formula Weight 72.11g/mol
Percent Purity ≥99.0%
Grade ReagentPlus
Chemical Name or Material Tetrahydrofuran, Contains 250 ppm BHT as inhibitor
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H302-H319-H335-H351
DOTInformation 3/PGII
EINECSNumber 203-726-8
RTECSNumber LU5950000