missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%, Solstice

Catalog No. 6001688
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001688 204 L Metal Drum
6001687 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001688 Supplier Solstice Supplier No. PP341204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

CAS Max % 100.0000%
Density 0.8892 g/cm3
Quantity 204 L
Flash Point −14°C
Packaging Metal Drum
UN Number UN2056
Water 0.05% max.
Beilstein 102391
Boiling Point 66°C
Solubility Information Miscible in Water at 20°C
Formula Weight 72.11
Percent Purity >99.9%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Tetrahydrofuran, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran