missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Optima™ for HPLC, Fisher Chemical™

Catalog No. p-216361
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Wide Neck Amber Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T4271 1 L Wide Neck Amber Bottle
T4274 4 L Amber Soda Lime Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T4271 Supplier Thermo Fisher Scientific Supplier No. FLT4271
Only null left

High purity solvent for HPLC

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Also meets ACS specifications
  • Intended for research use only

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

pH 7 to 8
Melting Point -108.4°C
Absorbance 1.0 AU max. at 210nm, 0.6 AU max. at 215nm, 0.3 AU max. at 230nm, 0.1 AU max. at 254nm
Color Colorless
Density 0.880g/cm³
Quantity 1 L
Identification Pass Test
Packaging Wide Neck Amber Bottle
Physical Form Liquid
Refractive Index 1.4040 to 1.4070 (at 25°C)
Residue after Evaporation 5ppm max.
Assay Percent Range 99.9% min by GC
Vapor Pressure 200mbar at 20°C
Viscosity 0.55 cP at 20°C
UN Number 2056
Water 0.02% max.
Boiling Point 66°C
Formula Weight 72.11
Percent Purity ≥99.9%
Grade High purity mobile phase for HPLC
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1
DOTInformation DOT Class 3, : Flammable Liquid