missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran Non UV, MilliporeSigma™
Supplier: Technidata TX02821
Specifications
| Tetrahydrofuran | |
| 109-99-9 | |
| 4 L | |
| 215°C | |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| C1CCOC1 | |
| 72.11 | |
| CHEBI:26911 |
| SeccoSolv™, Dried | |
| 0.89g/cm3 (20°C) | |
| 1.5 to 12.4%(V) | |
| Glass Bottle | |
| -CH2(CH2)3O- | |
| 173 hPa (20°C) | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 |
Chemical Identifiers
| 109-99-9 | |
| 72.11 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
Safety and Handling
Recommended Storage : Store at +15°C to +25°C