missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran Non UV, MilliporeSigma™

Catalog No. MTX02826
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MTX02826 1 L Glass Bottle
MTX02821 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MTX02826 Supplier Technidata Supplier No. TX02826

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Density 0.89g/cm3 (20°C)
Quantity 1 L
Explosion Limit 1.5 to 12.4%(V)
Ignition Point 215°C
Packaging Glass Bottle
Linear Formula -CH2(CH2)3O-
Vapor Pressure 173 hPa (20°C)
Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran, SeccoSolv™, Dried
Recommended Storage Store at +15°C to +25°C