missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran For spectroscopy Uvasol™, MilliporeSigma™

Catalog No. M1081102500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1081102500 2.5 L Glass Bottle
M1081100500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1081102500 Supplier Technidata Supplier No. 1.08110.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For spectroscopy Uvasol™

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

pH 7 to 8 (200g/L, H2O, (at 20°C)
Melting Point -108°C
Color Colorless
Density 0.89g/cm3 at 20°C
Quantity 2.5 L
Flash Point −20°C (−4°F)
Ignition Point 215°C (419°F)
Odor Ether-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 173 hPa (20°C)
UN Number 2056
Boiling Point 65°C to 66°C
Solubility Information Soluble in water at 20°C
Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H319: Causes serious eye irritation.
Precautionary Statement H335: May cause respiratory irritation.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protect from light.