missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran CHROMASOLV™ Plus, inhibitor-free, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

Catalog No. p-7073733
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001669 100 mL
6001670 1 L
6001671 2.5 L
6001675 6 x 1 L
6001673 4 x 2.5 L
6001674 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
May form explosive peroxides.
Catalog No. 6001669 Supplier Honeywell Chemicals Supplier No. 34865100ML
Only null left
May form explosive peroxides.

Suitable for HPLC, spectrophotometry, environmental testing.

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Absorbance absorption/300nm ≤0.02
Density 0.889g/mL at 25°C
Quantity 100 mL
Refractive Index n20/D 1.407
Linear Formula C4H8O
Vapor Pressure 143mmHg (20°C)
Beilstein 102391
Boiling Point 65°C to 67°C
Formula Weight 72.11g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material Tetrahydrofuran, inhibitor-free, for HPLC
Show More Show Less
Hazard Statement P210-P261-P281-P305 + P351 + P338
Precautionary Statement H225-H319-H335-H351
EINECSNumber 203-726-8
RTECSNumber LU5950000