missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran (Certified), Fisher Chemical™

Catalog No. T397500
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T397500 500 mL Glass Bottle
T3971 1 L Glass Bottle
T3974 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T397500 Supplier Thermo Fisher Scientific Supplier No. T397 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-99-9 , 128-37-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

pH 7 to 8
Melting Point -108.4°C
Color Colorless
Density 0.880g/cm³
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.884 to 0.887 (at 25°C)
Vapor Pressure 200mbar at 20°C
Viscosity 0.55 cP at 20°C
UN Number 2056
Water ≤0.02%
Boiling Point 66°C
Formula Weight 72.11
Grade Contains about 0.025% butylated hydroxytoluene as a preservative
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Safety handling Solvent
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1
DOTInformation DOT Class 3, : Flammable Liquid