missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Biograde, 99.8%, unstab.

Catalog No. p-7059392
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44505K2 1 L
AA44505K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44505K2 Supplier Thermo Scientific Chemicals Supplier No. 44505K2
Only null left

Nucleic acid and peptide synthesis | CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Color Colorless
Density 0.889 g/mL
Boiling Point 66°C
Quantity 1 L
Flash Point −17°C (1°F)
Odor Ether-like
Physical Form Liquid
Refractive Index 1.407
UN Number UN2056
Beilstein 102391
Merck Index 14,9211
Solubility Information Miscible with water.
Formula Weight 72.11
Percent Purity 99.8%
Grade Biograde
Chemical Name or Material Tetrahydrofuran, Unstabilized
Show More Show Less
Hazard Category H225-H302-H319-H335-H336-H351
Hazard Statement GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
EINECSNumber 203-726-8
RTECSNumber LU5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only