missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%, Honeywell Burdick & Jackson

Catalog No. p-7111835
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001679 1 L Glass Bottle
6001678 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001679 Supplier Honeywell Chemicals Supplier No. 3401L
Only null left

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Absorbance 0.18 AU max. at 250 nm, 0.02 AU max. at 300 nm, 0.005 AU max. at 350 nm, 0.005 AU max. at 400 nm
CAS Max % 100.0000%
Density 0.8892 g/cm3
Boiling Point 66°C
Quantity 1 L
Flash Point −14°C
Packaging Glass Bottle
UN Number UN2056
Beilstein 102391
Formula Weight 72.11
Percent Purity >99.9%
Grade HPLC Gradient
Chemical Name or Material Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran