missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%, Honeywell Burdick & Jackson
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.8892 g/cm3 |
| Quantity | 20 L |
| Flash Point | −14°C |
| Packaging | Returnable NOWPAK™ II |
| UN Number | UN2056 |
| Beilstein | 102391 |
| Boiling Point | 66°C |
| Formula Weight | 72.11 |
| Percent Purity | >99.9% |
| Show More |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran |