missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, for GPC, for GC, non-spectrophotometric grade, contains BHT, >99.9%, Solstice
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.8892 g/cm3 |
| Quantity | 200 L |
| Flash Point | −14°C |
| Packaging | Returnable Steel Drum |
| UN Number | UN2056 |
| Water | 0.01% max. |
| Beilstein | 102391 |
| Boiling Point | 66°C |
| Solubility Information | Miscible in Water at 20°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran |